Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

442515

Supelco

Chloroacetic acid

analytical standard

Synonym(s):

Monochloroacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2COOH
CAS Number:
Molecular Weight:
94.50
Beilstein/REAXYS Number:
605438
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

vapor density

3.26 (vs air)

vapor pressure

0.75 mmHg ( 20 °C)

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

189 °C (lit.)

mp

60-63 °C (lit.)

application(s)

agriculture
environmental

format

neat

storage temp.

room temp

SMILES string

OC(=O)CCl

InChI

1S/C2H3ClO2/c3-1-2(4)5/h1H2,(H,4,5)

InChI key

FOCAUTSVDIKZOP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

258.8 °F - closed cup

flash_point_c

126 °C - closed cup

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xuchun Li et al.
Environmental science & technology, 46(13), 7342-7349 (2012-06-12)
Most halogenated organic compounds (HOCs) are toxic and persistent, and their efficient destruction is currently a challenge. Here, we proposed a sulfite/UV (253.7 nm) process to eliminate HOCs. Monochloroacetic acid (MCAA) was selected as the target compound and was degraded
Patrick Levallois et al.
Epidemiology (Cambridge, Mass.), 23(2), 267-276 (2012-02-10)
There is concern about possible effects of disinfection by-products on reproductive outcomes. The purpose of this study was to evaluate the association between maternal exposure to chlorination by-products and the risk of delivering a small for-gestational-age (SGA) neonate. We conducted
Khalid Alhooshani et al.
Talanta, 86, 109-113 (2011-11-09)
For the first time, haloacetic acids and aromatic acetic acids were extracted from wastewater samples using electromembrane extraction (EME). A thin layer of toluene immobilized on the walls of a polypropylene membrane envelope served as an artificial supported liquid membrane
Wang Yanli et al.
Carbohydrate research, 344(13), 1764-1769 (2009-08-12)
Carboxymethyl starch (CMS) was obtained as a product of the reaction of starch and monochloroacetic acid (MCA) in the presence of sodium hydroxide. The influence of the molar ratio of NaOH/AGU, the molar ratio of MCA/AGU, the reaction time, reaction
Chun-Hung Chen et al.
Toxicology, 303, 72-82 (2012-10-30)
Chloroacetic acid (CA), a toxic chlorinated analog of acetic acid, is widely used in chemical industries as an herbicide, detergent, and disinfectant, and chemical intermediates that are formed during the synthesis of various products. In addition, CA has been found

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service