Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

442369

Supelco

2-Propanol

analytical standard

Synonym(s):

sec-Propyl alcohol, IPA, Isopropanol, Isopropyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHOH
CAS Number:
Molecular Weight:
60.10
Beilstein/REAXYS Number:
635639
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

analytical standard

vapor density

2.1 (vs air)

vapor pressure

33 mmHg ( 20 °C)
44 mmHg ( 25 °C)

autoignition temp.

750 °F

expl. lim.

2.0-12.7 %, 93 °C

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.377 (lit.)

bp

82 °C (lit.)

mp

−89.5 °C (lit.)

density

0.785 g/mL at 25 °C (lit.)

application(s)

environmental

SMILES string

CC(C)O

InChI

1S/C3H8O/c1-3(2)4/h3-4H,1-2H3

Inchi Key

KFZMGEQAYNKOFK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

target_organs

Central nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

53.6 °F - closed cup

flash_point_c

12.0 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lok Kumar Shrestha et al.
Journal of the American Chemical Society, 135(2), 586-589 (2013-01-02)
A new class of fullerene (C(60)) crystals with bimodal pore architectures consisting of macropores and mesopores was synthesized by using a liquid-liquid interfacial precipitation (LLIP) method involving an interface between isopropyl alcohol (IPA) and a saturated solution of C(60) in
Cristian Palmiere et al.
Forensic science international, 226(1-3), 54-61 (2013-01-15)
The aim of this study was to investigate the usefulness of postmortem biochemical investigations in the diagnosis of fatal hypothermia. 10 cases of fatal hypothermia and 30 control cases were selected. A series of biochemical parameters, such as glucose, acetone
Juergen Graessler et al.
PloS one, 4(7), e6261-e6261 (2009-07-16)
Dyslipoproteinemia, obesity and insulin resistance are integrative constituents of the metabolic syndrome and are major risk factors for hypertension. The objective of this study was to determine whether hypertension specifically affects the plasma lipidome independently and differently from the effects
Miranda Suchomel et al.
Infection control and hospital epidemiology, 34(3), 245-250 (2013-02-08)
The World Health Organization (WHO) has published "Guidelines on Hand Hygiene in Health Care" recommending 2 hand rub formulations based on 80% vol/vol ethanol or 75% vol/vol isopropanol for local production in healthcare settings where commercial products are not available
Jun Cao et al.
Electrophoresis, 34(2), 324-330 (2012-11-20)
A novel additive of multi-walled carbon nanotubes (MWNTs) dispersed with cationic surfactants or mixed cationic/anionic surfactants was used for MEEKC separation of eight phenolic compounds, four glycosides, and one phenanthraquinone. In this context, several parameters affecting MEEKC separation were studied

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service