Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

442260

Supelco

1-Chlorooctadecane

analytical standard

Synonym(s):

Octadecyl chloride, Stearyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)16CH2Cl
CAS Number:
Molecular Weight:
288.94
Beilstein/REAXYS Number:
1703350
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

analytical standard

CofA

current certificate can be downloaded

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.451 (lit.)

bp

157-158 °C/1.5 mmHg (lit.)

density

0.849 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

storage temp.

room temp

SMILES string

CCCCCCCCCCCCCCCCCCCl

InChI

1S/C18H37Cl/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19/h2-18H2,1H3

InChI key

VUQPJRPDRDVQMN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Product 442260 has been discontinued. Product 47584-U may be an acceptable replacement.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Aquatic Chronic 4 - Skin Irrit. 2

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

230.0 °F - closed cup

flash_point_c

110 °C - closed cup

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elisabeth Aubert et al.
Lipids, 39(1), 75-79 (2004-04-02)
The lipids of the gram-negative marine bacterium Marinobacter hydrocarbonoclasticus, cultivated in synthetic seawater supplemented with 1-chlorooctadecane as sole source of carbon, were isolated, purified, and their structures determined. Three pools of lipids were isolated according to the sequential procedure used:
G L Murphy et al.
Journal of bacteriology, 156(3), 1158-1164 (1983-12-01)
The cellular fatty acid composition of Mycobacterium vaccae JOB5 and Mycobacterium convolutum R22 was examined after growth on n-alkanes and compared with the fatty acids of the organisms after growth on 1-chlorohexadecane and 1-chlorooctadecane. Growth on n-alkanes resulted in normal
[Information from the Soviet Toxicology Center].
Gigiena truda i professional'nye zabolevaniia, (3)(3), 52-55 (1981-03-01)
Che C Colpitts et al.
Journal of virology, 88(14), 7806-7817 (2014-05-03)
Primary attachment to cellular glycans is a critical entry step for most human viruses. Some viruses, such as herpes simplex virus type 1 (HSV-1) and hepatitis C virus (HCV), bind to heparan sulfate, whereas others, such as influenza A virus
Becker Awqatty et al.
The Analyst, 139(20), 5230-5238 (2014-08-16)
The advanced functionality of portable devices such as smart phones provides the necessary hardware to potentially perform complex diagnostic measurements in any setting. Recent research and development have utilized cameras and data acquisition properties of smart phones to create diagnostic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service