Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

40092

Supelco

4,4′-DDD solution

certified reference material, 5000 μg/mL in methanol

Synonym(s):

1,1-Dichloro-2,2-bis(4-chlorophenyl)ethane

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Beilstein/REAXYS Number:
1914072
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:

grade

certified reference material
TraceCERT®

product line

TraceCERT®

packaging

ampule of 1 mL

concentration

5000 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture

format

single component solution

storage temp.

2-8°C

SMILES string

ClC(Cl)C(c1ccc(Cl)cc1)c2ccc(Cl)cc2

InChI

1S/C14H10Cl4/c15-11-5-1-9(2-6-11)13(14(17)18)10-3-7-12(16)8-4-10/h1-8,13-14H

InChI key

AHJKRLASYNVKDZ-UHFFFAOYSA-N

Gene Information

mouse ... Esr1(13982)
rat ... Ar(24208)

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

51.8 °F - closed cup

flash_point_c

11 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jiang-Ping Wu et al.
Environmental toxicology and chemistry, 31(4), 705-711 (2011-12-16)
Contamination with agricultural pesticides such as dichlorodiphenyltrichloroethane (DDT) and its metabolites, dichlorodiphenyldichloroethylene (DDE) and dichlorodiphenyldichloroethane (DDD), is among several proposed stressors contributing to the global declines in amphibian populations and species biodiversity. These chemicals were examined in insects and in
Mehmet Isleyen et al.
Journal of agricultural and food chemistry, 60(4), 1113-1121 (2012-01-10)
The grafting of melon plants onto cucurbit rootstocks is a common commercial practice in many parts of the world. However, certain cucurbits have been shown to accumulate large quantities of weathered persistent organic pollutants from the soil, and the potential
Mehmet Isleyen et al.
Environmental toxicology and chemistry, 31(8), 1699-1704 (2012-05-23)
Cucurbita pepo spp pepo (zucchini) is known as an exceptional weathered dichlorodiphenyldichloroethylene (DDE) accumulator, whereas Cucurbita pepo ssp ovifera (squash) is termed a nonaccumulator. Experiments were conducted with hybridized zucchini and squash to assess the inheritance pattern of DDX (the
C Witzig et al.
Heredity, 110(4), 389-397 (2013-01-10)
Prevention of malaria transmission throughout much of Africa is dependent on bednets that are impregnated with pyrethroid insecticides. Anopheles arabiensis is the major malaria vector in Chad and efforts to control this vector are threatened by the emergence of pyrethroid
Yvette Naudé et al.
Analytica chimica acta, 730, 112-119 (2012-05-29)
The organochlorine insecticide DDT (1,1,1-trichloro-2,2-bis(p-chlorophenyl)ethane) is still used for malaria vector control in certain areas of South Africa. The strict Stockholm Convention on Persistent Organic Pollutants (POPs) allows spraying on the inside of traditional dwellings with DDT. In rural villages

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service