Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

33009

Supelco

Dichlorodimethylsilane

for GC derivatization

Synonym(s):

DMDCS, Dimethyldichlorosilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2SiCl2
CAS Number:
Molecular Weight:
129.06
Beilstein/REAXYS Number:
605287
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

derivatization grade ((GC derivatization))
for GC derivatization

description

Silyl reagent

packaging

pkg of 100 mL

refractive index

n20/D 1.404 (lit.)

bp

70 °C (lit.)

mp

−76 °C (lit.)

density

1.07 g/mL at 25 °C (lit.)

compatibility

technique used for deactivating glassware and chromatographic supports

SMILES string

C[Si](C)(Cl)Cl

InChI

1S/C2H6Cl2Si/c1-5(2,3)4/h1-2H3

InChI key

LIKFHECYJZWXFJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dichlorodimethylsilane (DCDMS) is a hydrophobic in nature. It can be used to make ultra-water-repellent silica nanoparticle films and superhydrophobic γ-alumina membranes.[1]

Application

DCDMS in toluene was used for silanizing the vials overnight before performing solid-phase microextraction, for determination of polycyclic aromatic hydrocarbons in water samples.[2] It was also used as silanizing agent during a GC experiment for quantitative analysis of submicrogram amounts of the phenylthiohydantoins.[3]

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

33.8 °F - closed cup

flash_point_c

1 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R A Doong et al.
Journal of chromatography. A, 879(2), 177-188 (2000-07-13)
A solid-phase microextraction (SPME) procedure has been developed for the determination of 16 US Environmental Protection Agency promulgated polycyclic aromatic hydrocarbons (PAHs). Five kinds of SPME fibers were used and compared in this study. The extracted sample was analyzed by
Influences of dichlorodimethylsilane treatment on soil hydrophobicity, thermal conductivity, and electrical conductivity.
Ju, Zhaoqiang, Tusheng Ren, and Robert Horton
Soil Science, 173, 425-432 (2008)
Analysis of amino acid phenylthiohydantoins by gas chromatography.
J J Pisano et al.
The Journal of biological chemistry, 244(20), 5597-5607 (1969-10-25)
Wonmi Ahn et al.
ACS nano, 4(7), 4181-4189 (2010-06-23)
Uniform hexagonal arrays of diverse nanotemplated metal structures were formed via selective electroless gold plating on particle-lithographed dimethyldichlorosilane layers. Surface-associated water at silica bead interstices was shown to correlate with the formation of silane rings with outer ring diameters ranging
R Bos et al.
Microbiology (Reading, England), 142 ( Pt 9), 2355-2361 (1996-09-01)
Co-adhesion between oral microbial pairs (i.e. adhesion of a planktonic microorganism to a sessile organism adhering to a substratum surface) has been described as a highly specific interaction, mediated by stereochemical groups on the interacting microbial cell surfaces, and also

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service