Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

13390

Supelco

Styrene-DVB Copolymer

18-100 mesh, pkg of 1 kg

Synonym(s):

Poly(styrene-co-divinylbenzene), Styrene Divinylbenzene copolymer, Polystyrene crosslinked with divinylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH2CH(C6H5)]x[CH2CH[C6H4(CHCH2)]]y
CAS Number:
MDL number:
UNSPSC Code:
23151817
PubChem Substance ID:

description

Styrene-DVB

Quality Level

form

beads
slurry

packaging

pkg of 1 kg

manufacturer/tradename

Dow Chemical

technique(s)

LPLC: suitable
solid phase extraction (SPE): suitable

matrix

styrene/divinylbenzene gel

particle size

18-100 mesh

SMILES string

C=Cc1ccccc1.C=Cc2ccc(C=C)cc2.C=Cc3cccc(C=C)c3.C=Cc4ccccc4C=C

InChI

1S/C10H12.C10H10.C8H8/c2*1-3-9-7-5-6-8-10(9)4-2;1-2-8-6-4-3-5-7-8/h3,5-8H,1,4H2,2H3;3-8H,1-2H2;2-7H,1H2

InChI key

NWUYHJFMYQTDRP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Styrene is the simplest alkenylbenzene which is an unsaturated aromatic monomer.[1] It is widely used as column packing material in liquid chromatography for separation of both organic and inorganic ions.[2] Styrene-divinylbenzene Copolymer is used in inorganic trace analysis for solid phase extraction preconcentration.[3]

application

Styrene-divinylbenzene Copolymer is used as packing material (solid phase) to determine the contamination of groundwater by pesticides using solid phase extraction based method.[4]

pictograms

Health hazard

signalword

Warning

hcodes

Hazard Classifications

STOT RE 2

target_organs

hearing organs

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

  1. Which document(s) contains shelf-life or expiration date information for a given product?

    If available for a given product, the recommended re-test date or the expiration date can be found on the Certificate of Analysis.

  2. How do I get lot-specific information or a Certificate of Analysis?

    The lot specific COA document can be found by entering the lot number above under the "Documents" section.

  3. What is the particle size of Product 13390, Dow Styrene-divinylbenzene Copolymer?

    This Dow Styrene-DVB is listed at 18-100 mesh, which makes the particle size to be between 150-1000 microns.  The specifications do not include a mean or uniformity coefficient, so the bead size may be skewed from lot to lot.

  4. How do I find price and availability?

    There are several ways to find pricing and availability for our products. Once you log onto our website, you will find the price and availability displayed on the product detail page. You can contact any of our Customer Sales and Service offices to receive a quote.  USA customers:  1-800-325-3010 or view local office numbers.

  5. What is the Department of Transportation shipping information for this product?

    Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product. 

  6. My question is not addressed here, how can I contact Technical Service for assistance?

    Ask a Scientist here.

Sorption of magnesium-picolinate complexes on a macroporous styrene-divinylbenzene copolymer containing immobilized 8-hydroxyquinoline and on the copolymer itself.
Persaud, Gocool, and Frederick F. Cantwell.
Canadian Journal of Chemistry, 70, 926-930 (1992)
Determination of pesticides multiresidues in shallow groundwater in a cotton-growing region of Mato Grosso, Brazil
Leandro, Carbo, et al.
Journal of the Brazilian Chemical Society, 19, 1111-1117 (2008)
Dóra Marosvári et al.
Journal of neuropathology and experimental neurology, 77(4), 268-273 (2018-01-24)
The primary aim of this study was to determine mTOR-pathway activity in primary central nervous system lymphoma (PCNSL), which could be a potential target for therapy. After demonstrating that p-S6 positivity largely exceeded mTOR activity, we aimed to identify other
Styrene-divinyl benzene copolymers: synthesis, characterization, and their role in inorganic trace analysis
Rao, T. Prasada, R. S. Praveen, and Sobhi Daniel.
Critical Reviews in Analytical Chemistry, 34, 177-193 (2004)
Kerstin Hoppe et al.
Scientific reports, 6, 33372-33372 (2016-09-21)
Malignant hyperthermia (MH) is a pharmacogenetic disorder of skeletal muscle metabolism which is characterized by generalized muscle rigidity, increased body temperature, rhabdomyolysis, and severe metabolic acidosis. The underlying mechanism of MH involves excessive Ca(2+) release in myotubes via the ryanodine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service