Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V9514

Sigma-Aldrich

VGX-1027

≥98%

Synonym(s):

(S,R)-3-Phenyl-4,5-dihydro-5-isoxasole acetic acid, 2-Isoxazoline-5-acetic acid, 3-phenyl-, 5-Isoxazoleacetic acid, 4,5-dihydro-3-phenyl-, GIT27

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C11H11NO3
CAS Number:
Molecular Weight:
205.21
MDL number:
UNSPSC Code:
12352200

assay

≥98%

form

solid

color

white

solubility

DMSO: ≥30 mg/mL

Storage temp.

2-8°C

SMILES string

OC(=O)CC1CC(=NO1)c2ccccc2

Biochem/physiol Actions

VGX-1027 is a potent immunomodulatory compound.
VGX-1027 is a potent nitric oxide-donating immunomodulatory compound, which markedly decreased the cumulative incidence of diabetes and insulitis in NOD mice model with spontaneous or accelerated forms of diabetes induced either by injection of cyclophosphamide or by transfer of spleen cells from acutely diabetic syngeneic donors.

Pictograms

Health hazardExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Resp. Sens. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service