Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0312

Sigma-Aldrich

Acivicin

≥98% (HPLC)

Synonym(s):

(a-S, 5S)-a-Amino-3-chloro-4,5-dihydro-5-isoxazoleacetic acid, AT 125, Antibiotic AT 125, NSC 163501, U 42126

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H7ClN2O3
CAS Number:
Molecular Weight:
178.57
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

H2O: 10 mg/mL (warmed)

shipped in

dry ice

storage temp.

−20°C

SMILES string

N[C@@H]([C@@H]1CC(Cl)=NO1)C(O)=O

InChI

1S/C5H7ClN2O3/c6-3-1-2(11-8-3)4(7)5(9)10/h2,4H,1,7H2,(H,9,10)/t2-,4-/m0/s1

InChI key

QAWIHIJWNYOLBE-OKKQSCSOSA-N

Biochem/physiol Actions

Acivicin is an antibiotic that inhibits g-glutamyl transpeptidase and transmembrane glutathione transport. Acivicin is a potent antitumor agent that induces apoptosis in human lymphoblastoid cells.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christianne Bandeira-Melo et al.
The Journal of allergy and clinical immunology, 109(6), 975-979 (2002-06-14)
Eosinophils contain preformed stores of IL-4 within their cytoplasmic granules, but physiologic stimuli to release IL-4 from eosinophils are not yet defined. We evaluated whether cysteinyl leukotrienes (CysLTs) could elicit IL-4 release from eosinophils. We used a dual-antibody capture and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service