Key Documents
SIAL0659
Sigma-Aldrich
Sigma® cryogenic vial
capacity 2 mL, External thread, sterile; irradiated (Radiation Sterilization. Sterility Assurance Level: SAL 10^5), pack of 500 ea (10 x 50 ea)
About This Item
UNSPSC Code:
41121704
NACRES:
NB.22
material
colorless tube
polypropylene
polypropylene tube
round bottom (self-standing)
sterility
sterile; irradiated (Radiation Sterilization. Sterility Assurance Level: SAL 10^5)
feature
graduated
External thread
self standing
packaging
pack of 10 × 50 ea
pack of 500 ea (10 x 50 ea)
manufacturer/tradename
Corning
Sigma-Aldrich
parameter
-196 °C min. temp.
capacity
2 mL
General description
  • Cryogenic vials are manufactured from polypropylene to withstand temperatures to -196 °C.
  • Features a silicone washer for a secure seal.
  • Can be color-coded with inserts.
  • All self-standing vials have a special base design allowing them to be locked into the cryogenic rack and tray, for single-handed manipulation.
Legal Information
Sigma is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)