Key Documents
SIAL0599
Sigma-Aldrich
Sigma® cell culture dish
diam. × H 150 mm × 25 mm, tissue-culture treated, sterile, pack of 12 × 5 ea
About This Item
UNSPSC Code:
41121812
sterility
sterile
packaging
pack of 12 × 5 ea
diam. × H
150 mm × 25 mm
surface area
148 cm2
binding type
Tissue Culture (TC)-treated surface
General description
  • Manufactured from optically-clear virgin polystyrene
  • Treated for optimal cell attachment
  • Sterilized by gamma radiation and certified nonpyrogenic
  • Have stacking beads to aid in handling
  • Lid supplied with vents to provide consistent gas exchange
Legal Information
Sigma is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)