Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SIAL0548

Sigma-Aldrich

Sigma® cell culture plate

size 48 wells, flat bottom, sterile, tissue-culture treated, pack of 100 ea individually packed

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122107
NACRES:
NB.22

material

clear polystyrene plate
clear wells
flat bottom

description

growth area 0.95 cm2
with lid

sterility

sterile

packaging

pack of 100 ea individually packed

size

48 wells

well diam.

11.0 mm

color

clear

binding type

Tissue Culture (TC)-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

  • Non-reversible lids with condensation rings to reduce contamination
  • Individual alphanumerical codes for well identification
  • Uniform footprint for ease in stacking
  • Treated for optimal cell attachment
  • Sterilized by gamma irradiation
  • Certified nonpyrogenic

Legal Information

Sigma is a registered trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service