Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

R5504

Sigma-Aldrich

Raney® nickel

50% in water, pH >9 (slurry)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

concentration

50% in water, pH >9 (slurry)

Application

Activated catalyst

Linkage

Analogous to Raney 28 or W-2.

Legal Information

Raney is a registered trademark of W.R. Grace & Co.-Conn.

replaced by

signalword

Danger

Hazard Classifications

Carc. 2 - Pyr. Liq. 1 - Skin Sens. 1 - STOT RE 1

Storage Class

4.2 - Pyrophoric and self-heating hazardous materials

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service