Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

O3752

Sigma-Aldrich

Orphenadrine hydrochloride

Synonym(s):

β-Dimethylaminoethyl 2-methylbenzhydryl ether hydrochloride, N,N-Dimethyl-2-(2-methylbenzhydryloxy)ethylamine hydrochloride, o-Methyldiphenhydramine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H23NO · HCl
CAS Number:
Molecular Weight:
305.84
Beilstein/REAXYS Number:
3745818
EC Number:
MDL number:
UNSPSC Code:
12352200

storage temp.

2-8°C

SMILES string

Cl.CN(C)CCOC(c1ccccc1)c2ccccc2C

InChI

1S/C18H23NO.ClH/c1-15-9-7-8-12-17(15)18(20-14-13-19(2)3)16-10-5-4-6-11-16;/h4-12,18H,13-14H2,1-3H3;1H

InChI key

UQZKYYIKWZOKKD-UHFFFAOYSA-N

Gene Information

human ... HRH1(3269)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Muscarinic receptor antagonist; H1 histamine receptor antagonist; muscle relaxant. Orphenadrine has also been reported to inhibit the noradrenergic transporter and to block the NMDA receptor ion channel.

replaced by

Product No.
Description
Pricing

pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marianna Borsodi et al.
Orvosi hetilap, 149(39), 1847-1852 (2008-09-23)
The authors compared the potency, safety and tolerability of combined infusion containing non-steroid anti-inflammation diclofenac and central muscle relaxant orphenadrine, and those of tramadol HCl, during postoperative pain relief after low and middle category operations. The test was an open
Bismi Edwin et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 97, 838-846 (2012-08-21)
Vibrational spectral analysis and quantum chemical computations based on density functional theory have been performed on the anti-neuro-degenerative drug Orphenadrine hydrochloride. The geometry, intermolecular hydrogen bond, and harmonic vibrational frequencies of the title molecule have been investigated with the help
Konrad Rejdak et al.
Brain research bulletin, 84(6), 389-393 (2011-01-29)
The current study was aimed to assess the convulsant potency of orphenadrine (ORPH) in rats together with a screen of different conventional antiepileptic drugs (AEDs) on their efficacy to suppress it. ORPH was administered intraperitoneally (i.p.) in doses of 50-80
Eberhard P Scholz et al.
Naunyn-Schmiedeberg's archives of pharmacology, 376(4), 275-284 (2007-10-30)
The anticholinergic antiparkinson drug orphenadrine is an antagonist at central and peripheral muscarinic receptors. Orphenadrine intake has recently been linked to QT prolongation and Torsade-de-Pointes tachycardia. So far, inhibitory effects on I (Kr) or cloned HERG channels have not been
Jean-François Desaphy et al.
Pain, 142(3), 225-235 (2009-02-17)
Orphenadrine is a drug acting on multiple targets, including muscarinic, histaminic, and NMDA receptors. It is used in the treatment of Parkinson's disease and in musculoskeletal disorders. It is also used as an analgesic, although its mechanism of action is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service