Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M7154

Sigma-Aldrich

2-Mercaptoethanol

suitable for electrophoresis

Synonym(s):

β-Mercaptoethanol, 2-Hydroxyethylmercaptan, BME, Thioethylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HSCH2CH2OH
CAS Number:
Molecular Weight:
78.13
Beilstein/REAXYS Number:
773648
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

vapor density

2.69 (vs air)

vapor pressure

1 mmHg ( 20 °C)

assay

>99% (GC)

expl. lim.

18 %

concentration

14.3 M (pure liquid)

technique(s)

electrophoresis: suitable

refractive index

n20/D 1.500 (lit.)

bp

157 °C (lit.)

solubility

water: soluble

density

1.114 g/mL at 25 °C (lit.)

cation traces

Cu: <1 ppm
Fe: <1 ppm
Pb: <5 ppm
Zn: <1 ppm

SMILES string

OCCS

InChI

1S/C2H6OS/c3-1-2-4/h3-4H,1-2H2

InChI key

DGVVWUTYPXICAM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

BME is suitable for reducing protein disulfide bonds prior to polyacrylamide gel electrophoresis and is usually included in a sample buffer for SDS-PAGE at a concentration of 5%. Cleaving intermolecular (between subunits) disulfide bonds allows the subunits of a protein to separate independently on SDS-PAGE. Cleaving intramolecular (within subunit) disulfide bonds allows the subunits to become completely denatured so that each peptide migrates according to its chain length with no influence due to secondary structure.

signalword

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Repr. 2 - Skin Irrit. 2 - Skin Sens. 1A - STOT RE 2 Oral

target_organs

Liver,Heart

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

165.2 °F - closed cup

flash_point_c

74 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Leon Tribolet et al.
The Journal of infectious diseases, 211(3), 416-425 (2014-08-21)
Na-ASP-2 is an efficacious hookworm vaccine antigen. However, despite elucidation of its crystal structure and studies addressing its immunobiology, the function of Na-ASP-2 has remained elusive. We probed a 9000-protein human proteome microarray with Na-ASP-2 and showed binding to CD79A
Katharina Rothe et al.
Blood, 123(23), 3622-3634 (2014-04-24)
Previous studies demonstrated that imatinib mesylate (IM) induces autophagy in chronic myeloid leukemia (CML) and that this process is critical to cell survival upon therapy. However, it is not known if the autophagic process differs at basal levels between CML
E M Michalak et al.
Cell death and differentiation, 15(6), 1019-1029 (2008-02-09)
The ability of p53 to induce apoptosis in cells with damaged DNA is thought to contribute greatly to its tumour suppressor function. P53 has been proposed to induce apoptosis via numerous transcriptional targets or even by direct cytoplasmic action. Two
Emily Cunningham Williams et al.
Human molecular genetics, 23(11), 2968-2980 (2014-01-15)
The disease mechanism of Rett syndrome (RTT) is not well understood. Studies in RTT mouse models have suggested a non-cell-autonomous role for astrocytes in RTT pathogenesis. However, it is not clear whether this is also true for human RTT astrocytes.
Nobuko Akiyama et al.
The Journal of experimental medicine, 211(12), 2425-2438 (2014-11-12)
Medullary thymic epithelial cells (mTECs) expressing the autoimmune regulator AIRE and various tissue-specific antigens (TSAs) are critical for preventing the onset of autoimmunity and may attenuate tumor immunity. However, molecular mechanisms controlling mTEC development remain elusive. Here, we describe the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service