Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

L0134

Sigma-Aldrich

β-Lipotropin Fragment 88-91

≥95% (HPLC)

Synonym(s):

Lys-Lys-Gly-Glu, MPF

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H36N6O7
CAS Number:
Molecular Weight:
460.53
MDL number:
UNSPSC Code:
12352200

assay

≥95% (HPLC)

UniProt accession no.

storage temp.

−20°C

SMILES string

NCCCCC(N)C(=O)NC(CCCCN)C(=O)NCC(=O)NC(CCC(O)=O)C(O)=O

Gene Information

human ... POMC(5443)

Biochem/physiol Actions

Melanotropin potentiating factor which gives a 5-fold enhancement of α-MSH activity at 0.5 nM

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Melanotropin potentiating factor is the C-terminal tetrapeptide of human beta-lipotropin.
R J Carter et al.
Nature, 279(5708), 74-75 (1979-05-03)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service