Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE17-0623-01

PhastGel® Homogeneous Pre-cast Gels

12.5% polyacrylamide, pack of 10 gels

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105300
NACRES:
NB.22

form

solid

packaging

pack of 10 gels

manufacturer/tradename

Cytiva 17062301

parameter

80-230 separation range

size

50 mm × 50 mm × 0.45 mm

color

Colorless

solubility

cold water: soluble (Insoluble in cold and hot water)

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Legal Information

PhastGel is a registered trademark of Cytiva

pictograms

Health hazard

signalword

Danger

hcodes

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service