G0900
Gramicidin S hydrochloride from Bacillus brevis (Nagano)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352200
Recommended Products
storage temp.
2-8°C
Application
A cyclic decapeptide antibiotic.
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
MULTIPLICITY OF THE CELLULOLYTIC SYSTEM OF TRICHODERMA VIRIDE II.
S SHIKATA
Journal of biochemistry, 56, 1-5 (1964-07-01)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service