Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C8612

Sigma-Aldrich

Chloral

anhydrous

Synonym(s):

Trichloroacetaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl3CCHO
CAS Number:
Molecular Weight:
147.39
Beilstein/REAXYS Number:
506422
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

anhydrous

assay

≥97% (GC)

drug control

USDEA Schedule IV

density

1.51 g/mL at 20 °C (lit.)

storage temp.

2-8°C

SMILES string

ClC(Cl)(Cl)C=O

Looking for similar products? Visit Product Comparison Guide

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation

target_organs

Lungs,Adrenal gland

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

167.0 °F - closed cup

flash_point_c

75 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service