Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

B6655

Sigma-Aldrich

3-tert-Butyl-4-hydroxyanisole

Synonym(s):

2-tert-Butyl-4-methoxyphenol, 3-BHA, BHA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H16O2
CAS Number:
Molecular Weight:
180.24
Beilstein/REAXYS Number:
1867499
EC Number:
MDL number:
UNSPSC Code:
12352100

SMILES string

COc1ccc(O)c(c1)C(C)(C)C

InChI

1S/C11H16O2/c1-11(2,3)9-7-8(13-4)5-6-10(9)12/h5-7,12H,1-4H3

InChI key

MRBKEAMVRSLQPH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Minimum 90% 3-BHA and maximum 8% 2-BHA

replaced by

Product No.
Description
Pricing

pictograms

Environment

hcodes

Hazard Classifications

Aquatic Chronic 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service