Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AV50255

Sigma-Aldrich

Anti-ND6 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-MTND6, Anti-NADH dehydrogenase, subunit 6 (complex I)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

mol wt

19 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

storage temp.

−20°C

Gene Information

human ... ND6(4541)

Immunogen

The immunogen for anti-ND6 antibody: synthetic peptide derected towards the of human ND6

Sequence

Synthetic peptide located within the following region: DGVVVVVNFNSVGSWMIYEGEGSGLIREDPIGAGALYDYGRWLVVVTGWT

Physical form

Lyophilized from PBS buffer with 2% sucrose

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Ravn et al.
European journal of human genetics : EJHG, 9(10), 805-809 (2002-01-10)
We report a novel point mutation in the gene for the mitochondrially encoded ND6 subunit of the NADH:ubiquinone oxidoreductase (complex I of the respiratory chain) in a patient with MELAS syndrome. The mutation causes a change from alanine to valine
Juhua Yang et al.
Mitochondrion, 9(4), 273-278 (2009-04-28)
We report the clinical and genetic characterization of a Chinese Leber's hereditary optic neuropathy (LHON) family with complete penetrance and high percentage of recovery. Sequence analysis of the complete mitochondrial DNA revealed the presence of heteroplasmic ND6/T14484C mutation and 27
Yulia V Surovtseva et al.
Proceedings of the National Academy of Sciences of the United States of America, 108(44), 17921-17926 (2011-10-18)
Basal transcription of human mitochondrial DNA (mtDNA) in vitro requires the single-subunit, bacteriophage-related RNA polymerase, POLRMT, and transcription factor h-mtTFB2. This two-component system is activated differentially at mtDNA promoters by human mitochondrial transcription factor A (h-mtTFA). Mitochondrial ribosomal protein L7/L12

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service