Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A6930

Sigma-Aldrich

β-N-Acetylhexosaminidase

recombinant, expressed in E. coli, buffered aqueous solution

Synonym(s):

β-N-Acetyl-D-hexosaminide N-acetyl-hexosaminohydrolase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204

recombinant

expressed in E. coli

form

buffered aqueous solution

UniProt accession no.

foreign activity

α-Glucosidase <1%
α-mannosidase, β-hexosaminidase, α-fucosidase, neuraminidase, and proteases, none detected (enzyme is expressed in glycosidase-free host)
β-Galactosidase <2%

storage temp.

2-8°C

Gene Information

human ... HEXA(3073)

Biochem/physiol Actions

Active on N-acetyl-β-D-glucosaminides and N-acetyl-β-D-galactosaminides.

Unit Definition

One unit will hydrolyze 1.0 μmole of p-nitrophenyl N-acetyl-β-D-glucosaminide to p-nitrophenol and N-acetyl-D-glucosamine per min at pH 5.0 at 37°C.

Physical form

Solution in 20 mM Tris-HCl, pH 7.5, 25 mM NaCl, 10 mM DTT

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tyler Mark Pierson et al.
Molecular genetics and metabolism, 108(1), 65-69 (2012-11-20)
A 12 year-old female presented with a seven-year history of progressive muscle weakness, atrophy, tremor and fasciculations. Cognition was normal. Rectal biopsy revealed intracellular storage material and biochemical testing indicated low hexosaminidase activity consistent with juvenile-onset G(M2)-gangliosidosis. Genetic evaluation revealed
Federica Vernuccio et al.
Recenti progressi in medicina, 103(12), 559-563 (2012-12-22)
Cardiorenal syndrome is a pathophysiological heart and kidney disorder, in which acute or chronic dysfunction of one organ induces a damage in the other. It's a syndrome more and more often encountered in clinical practice and this implies the need
A M Dygai et al.
Bulletin of experimental biology and medicine, 154(2), 278-282 (2013-01-19)
We studied the effect of mobilization of bone marrow multipotent stem cells induced by intragastric administration of pegylated hyaluronate-endo-β-N-acetylhexosaminidase (Peg-HEAHA) on hemopoiesis under conditions of experimental chronic hepatitis. Peg-HEAHA increased the counts of hemopoietic precursors in the hemopoietic tissue against
David J Vocadlo
Current opinion in chemical biology, 16(5-6), 488-497 (2012-11-14)
The addition of N-acetylglucosamine (GlcNAc) O-linked to serine and threonine residues of proteins is known as O-GlcNAc. This post-translational modification is found within multicellular eukaryotes on hundreds of nuclear and cytoplasmic proteins. O-GlcNAc transferase (OGT) installs O-GlcNAc onto target proteins
Motoshi Ouchi et al.
Journal of clinical laboratory analysis, 26(6), 473-480 (2012-11-13)
Urinary N-acetyl-β-D-glucosaminidase (NAG) excretion is increased in patients with impaired glucose tolerance (IGT). This study investigated when during the oral glucose tolerance test (OGTT) the plasma glucose, urine glucose, and insulin levels correlate most strongly with urinary N-acetyl-β-d-glucosaminidase (NAG) levels

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service