X0754
D-Xylulose 5-phosphate sodium salt
≥90% (TLC)
Synonym(s):
Xu-5-P
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
assay
≥90% (TLC)
storage temp.
−20°C
Looking for similar products? Visit Product Comparison Guide
Biochem/physiol Actions
Xylulose-5-phoshate (Xu-5-P) is a metabolite of glucose via the pentose phosphate pathway. In the presence of excess glucose, liver Xu-5-P may act as a signaling molecule that regulates the nuclear transport of carbohydrate response element binding protein (ChREBP), a transcription factor that induces the expression of enzymes involved in glucose metabolism and lipogenesis.
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service