V0375
DL-Valine
≥97% (TLC)
Synonym(s):
(±)-α-Aminoisovaleric acid, DL-2-Amino-3-methylbutanoic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
(CH3)2CHCH(NH2)COOH
CAS Number:
Molecular Weight:
117.15
Beilstein/REAXYS Number:
506689
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Recommended Products
assay
≥97% (TLC)
form
powder
color
white
mp
295 °C (dec.) (lit.)
SMILES string
CC(C)C(N)C(O)=O
InChI
1S/C5H11NO2/c1-3(2)4(6)5(7)8/h3-4H,6H2,1-2H3,(H,7,8)
InChI key
KZSNJWFQEVHDMF-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Yan-Mei Ma et al.
Fish & shellfish immunology, 45(2), 414-421 (2015-05-11)
Streptococcus iniae seriously affects the intensive farming of tilapias. Much work has been conducted on prevention and control of S. iniae infection, but little published information on the metabolic response is available in tilapias against the bacterial infection, and no metabolic
K Aaboe et al.
Diabetes, obesity & metabolism, 17(1), 74-81 (2014-09-23)
To examine whether 12 weeks of treatment with a dipeptidyl peptidase-4 (DPP-4) inhibitor, sitagliptin, influences the insulin secretion induced by glucose, glucose-dependent insulinotropic polypeptide (GIP) and glucagon-like peptide-1 (GLP-1) during a hyperglycaemic clamp in patients with type 2 diabetes (T2DM).
Aleksander Hinek et al.
Journal of dermatological science, 75(3), 173-182 (2014-07-13)
Vitamin C (L-ascorbic acid), a known enhancer of collagen deposition, has also been identified as an inhibitor of elastogenesis. Present studies explored whether and how the L-ascorbic acid derivative (+) sodium L-ascorbate (SA) would affect production of collagen and elastic
Yuka Kamei et al.
The Journal of biological chemistry, 289(46), 32081-32093 (2014-10-09)
Age-related damage accumulates and a variety of biological activities and functions deteriorate in senescent cells. However, little is known about when cellular aging behaviors begin and what cellular aging processes change. Previous research demonstrated age-related mRNA changes in budding yeast
Thomas Courivaud et al.
The Journal of biological chemistry, 290(34), 21007-21018 (2015-07-15)
Although E3 ubiquitin ligases are deemed to play key roles in normal cell function and homeostasis, whether their alterations contribute to cancer pathogenesis remains unclear. In this study, we sought to investigate potential mechanisms that govern WWP1/Tiul1 (WWP1) ubiquitin ligase
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service