Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T8403

Sigma-Aldrich

D-α-(3-Thienyl)glycine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H7NO2S
CAS Number:
Molecular Weight:
157.19
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

SMILES string

N[C@H](C(O)=O)c1ccsc1

InChI

1S/C6H7NO2S/c7-5(6(8)9)4-1-2-10-3-4/h1-3,5H,7H2,(H,8,9)/t5-/m0/s1

InChI key

BVGBBSAQOQTNGF-YFKPBYRVSA-N

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service