Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T8400

Sigma-Aldrich

Thr-Leu

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H20N2O4
CAS Number:
Molecular Weight:
232.28
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Storage temp.

−20°C

SMILES string

CC(C)CC(NC(=O)C(N)C(C)O)C(O)=O


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yi-Ying Lee et al.
Journal of bacteriology, 197(1), 159-173 (2014-10-22)
Proteus mirabilis is a dimorphic motile bacterium well known for its flagellum-dependent swarming motility over surfaces. In liquid, P. mirabilis cells are 1.5- to 2.0-μm swimmer cells with 4 to 6 flagella. When P. mirabilis encounters a solid surface, where
Naoko Okai et al.
AMB Express, 4, 20-20 (2014-06-21)
Gamma-aminobutyric acid (GABA), a building block of the biodegradable plastic polyamide 4, is synthesized from glucose by Corynebacterium glutamicum that expresses Escherichia coli glutamate decarboxylase (GAD) B encoded by gadB. This strain was engineered to produce GABA more efficiently from
Jun Zhang et al.
Applied and environmental microbiology, 81(8), 2852-2860 (2015-02-15)
Inorganic arsenic (As) is highly toxic and ubiquitous in the environment. Inorganic As can be transformed by microbial methylation, which constitutes an important part of the As biogeochemical cycle. In this study, we investigated As biotransformation by Pseudomonas alcaligenes NBRC14159.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service