Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T7258

Sigma-Aldrich

Trizma® oxalate

reagent grade

Synonym(s):

Di(tris[hydroxymethyl]aminomethane) oxalate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H24N2O10
CAS Number:
Molecular Weight:
332.30
MDL number:
UNSPSC Code:
12161700

grade

reagent grade

SMILES string

OC(=O)C(O)=O.NC(CO)(CO)CO.NC(CO)(CO)CO

Legal Information

Trizma is a registered trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service