Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T6771

Sigma-Aldrich

Triamterene

Synonym(s):

2,4,7-Triamino-6-phenylpteridine, 6-Phenyl-2,4,7-pteridinetriamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H11N7
CAS Number:
Molecular Weight:
253.26
EC Number:
MDL number:
UNSPSC Code:
12352200
Product T6771 is not currently sold in your country. Contact Technical Service

storage temp.

2-8°C

SMILES string

Nc1nc(N)c2nc(-c3ccccc3)c(N)nc2n1

InChI

1S/C12H11N7/c13-9-7(6-4-2-1-3-5-6)16-8-10(14)18-12(15)19-11(8)17-9/h1-5H,(H6,13,14,15,17,18,19)

InChI key

FNYLWPVRPXGIIP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

51.8 °F - closed cup

flash_point_c

11 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service