Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T6633

Sigma-Aldrich

Thymidylyl(3′→5′)thymidylyl(3′→5′)-2′-deoxycytidine disodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C29H37N7O18P2Na2
CAS Number:
Molecular Weight:
879.57
MDL number:
UNSPSC Code:
12352204

storage temp.

−20°C

SMILES string

[Na].CC1=CN(C2CC(OP(O)(=O)OCC3OC(CC3OP(O)(=O)OCC4OC(CC4O)N5C=CC(N)=NC5=O)N6C=C(C)C(=O)NC6=O)C(CO)O2)C(=O)NC1=O

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service