T6147
Testosterone
suitable for cell culture
Synonym(s):
17β-Hydroxy-3-oxo-4-androstene, 17β-Hydroxy-4-androsten-3-one, 4-Androsten-17β-ol-3-one, trans-Testosterone
About This Item
Recommended Products
drug control
USDEA Schedule III; Home Office Schedule 4.2; regulated under CDSA - not available from Sigma-Aldrich Canada
technique(s)
cell culture | mammalian: suitable
SMILES string
C[C@]12CC[C@H]3[C@@H](CCC4=CC(=O)CC[C@]34C)[C@@H]1CC[C@@H]2O
InChI
1S/C19H28O2/c1-18-9-7-13(20)11-12(18)3-4-14-15-5-6-17(21)19(15,2)10-8-16(14)18/h11,14-17,21H,3-10H2,1-2H3/t14-,15-,16-,17-,18-,19-/m0/s1
InChI key
MUMGGOZAMZWBJJ-DYKIIFRCSA-N
Gene Information
human ... AR(367)
Looking for similar products? Visit Product Comparison Guide
Biochem/physiol Actions
comparable product
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Acute 1 - Carc. 2 - Repr. 1B
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type P3 (EN 143) respirator cartridges
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service