Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T5767

Sigma-Aldrich

N,N,N′,N′-Tetramethylguanidine

Synonym(s):

1,1,3,3-Tetramethylguanidine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NC(=NH)N(CH3)2
CAS Number:
Molecular Weight:
115.18
Beilstein/REAXYS Number:
969608
EC Number:
MDL number:
UNSPSC Code:
12352200

bp

162-163 °C (lit.)

density

0.916 g/mL at 20 °C (lit.)

storage temp.

2-8°C

SMILES string

CN(C)C(=N)N(C)C

InChI

1S/C5H13N3/c1-7(2)5(6)8(3)4/h6H,1-4H3

InChI key

KYVBNYUBXIEUFW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Met. Corr. 1 - Skin Corr. 1B

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

122.0 °F - closed cup

flash_point_c

50 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service