T4634
4-Thiouridine 5′-monophosphate sodium salt
~98%
Synonym(s):
4-Thio UMP
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C9H13N2O8PS
CAS Number:
Molecular Weight:
340.25
MDL number:
UNSPSC Code:
12352204
assay
~98%
impurities
≤2% ammonia
storage temp.
−20°C
SMILES string
OC1C(O)C(OC1COP(O)(O)=O)N2C=CC(=S)NC2=O
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service