Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T3902

Sigma-Aldrich

Thiamine hydrochloride

suitable for plant cell culture, BioReagent

Synonym(s):

Aneurine hydrochloride, Vitamin B1 hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H17ClN4OS · HCl
CAS Number:
Molecular Weight:
337.27
Beilstein/REAXYS Number:
3851771
EC Number:
MDL number:
UNSPSC Code:
12352200
eCl@ss:
34058006
PubChem Substance ID:

product line

BioReagent

technique(s)

cell culture | plant: suitable

mp

250 °C (dec.) (lit.)

application(s)

agriculture

SMILES string

CC1=NC(N)=C(C[N+]2=CSC(CCO)=C2C)C=N1.Cl.[Cl-]

InChI

1S/C12H17N4OS.2ClH/c1-8-11(3-4-17)18-7-16(8)6-10-5-14-9(2)15-12(10)13;;/h5,7,17H,3-4,6H2,1-2H3,(H2,13,14,15);2*1H/q+1;;/p-1

Inchi Key

DPJRMOMPQZCRJU-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Thiamine is a precursor of thiamine diphosphate, a coenzyme for enzymes that catalyse two-carbon unit transfer reactions by enzymes such as pyruvate dehydrogenase and 2-oxoglutarate dehydrogenase; branched-chain α-keto acid dehydrogenase; 2-hydroxyphytanoyl-CoA lyase and transketolase. Thiamine is added to a variety of plant cell cultures to enhance processes such as shoot formation.

Pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Baruah et al.
Indian journal of experimental biology, 39(9), 947-949 (2002-02-08)
A protocol was developed for high frequency plant regeneration in H. patulum by shoot-tip culture. H. patulum plants were collected from a wild source growing at high altitude in the eastern Himalayas. Multiple buds were initiated from shoot-tips cultured on
M Ramesh et al.
Indian journal of experimental biology, 47(1), 68-74 (2009-03-26)
A reliable and reproducible protocol has been developed for high frequency plant regeneration from 4-5 mm long leaf base segments of 4 days old in vitro germinated seedlings of indica rice (Oryza sativa) cultivar Rasi. The effect of age of
Joseph E Kerschner et al.
Archives of otolaryngology--head & neck surgery, 135(1), 33-39 (2009-01-21)
To investigate genetic differences in middle ear mucosa (MEM) with nontypeable Haemophilus influenzae (NTHi) infection. Genetic upregulation and downregulation occurs in MEM during otitis media (OM) pathogenesis. A comprehensive assessment of these genetic differences using the techniques of complementary DNA
Samir Ahmed El-Shazly et al.
International journal of immunopathology and pharmacology, 28(2), 227-239 (2015-03-31)
Diabetes mellitus (DM) is a chronic metabolic disorder characterized by hyperglycemia due to abnormalities in either insulin secretion or action. A range of vanadium complexes have been synthesized and demonstrated to be effective in lowering hyperglycemia. Thiamine administration was also
Ashish K Singh et al.
Alcoholism, clinical and experimental research, 38(7), 2127-2137 (2014-06-17)
Chronic ethanol (EtOH) consumption is associated with oxidative tissue damage, decrease in antioxidant enzyme activities, and increase in hepatic and plasma lipids. This study investigates the effect of modified probiotic Escherichia coli Nissle 1917 (EcN) secreting pyrroloquinoline quinone (PQQ) against

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service