T2408
Tolazamide
Synonym(s):
1-(Hexahydro-1H-azepin-1-yl)-3-(p-toluenesulfonyl)urea
About This Item
solubility
0.1 M NaOH: soluble 4.3 mg/ml
45% (w/v) aq 2-hydroxypropyl-β-cyclodextrin: soluble 2.7 mg/ml
DMSO: soluble 32.5 mg/ml
acetone: soluble 50 mg/ml, clear, colorless to light yellow
SMILES string
Cc1ccc(cc1)S(=O)(=O)NC(=O)NN2CCCCCC2
InChI
1S/C14H21N3O3S/c1-12-6-8-13(9-7-12)21(19,20)16-14(18)15-17-10-4-2-3-5-11-17/h6-9H,2-5,10-11H2,1H3,(H2,15,16,18)
InChI key
OUDSBRTVNLOZBN-UHFFFAOYSA-N
Gene Information
human ... ALB(213) , KCNJ1(3758)
Looking for similar products? Visit Product Comparison Guide
Application
Biochem/physiol Actions
Preparation Note
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service