Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T162

Supelco

Testosterone decanoate

Synonym(s):

17β-Hydroxy-4-androsten-3-one 17-decanoate, 4-Androsten-17β-ol-3-one 17-caprate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H46O3
CAS Number:
Molecular Weight:
442.67
EC Number:
MDL number:
UNSPSC Code:
41116107

drug control

USDEA Schedule III; Home Office Schedule 4.2; regulated under CDSA - not available from Sigma-Aldrich Canada

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

SMILES string

CCCCCCCCCC(=O)O[C@H]1CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C

InChI

1S/C29H46O3/c1-4-5-6-7-8-9-10-11-27(31)32-26-15-14-24-23-13-12-21-20-22(30)16-18-28(21,2)25(23)17-19-29(24,26)3/h20,23-26H,4-19H2,1-3H3/t23-,24-,25-,26-,28-,29-/m0/s1

InChI key

LBERVHLCXUMDOT-MPZZESAYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Carc. 2 - Repr. 1B

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R W Hamill et al.
Brain research, 299(2), 331-337 (1984-05-14)
The effects of the hormone testosterone on neurotransmitter synthesis in peripheral sympathetic ganglia were examined in adult male Sprague-Dawley rats. Tyrosine hydroxylase (T-OH), the rate limiting enzyme in catecholamine biosynthesis was examined in the hypogastric (HG), coeliac (CG), and superior
J C do Prado et al.
Parasitology research, 85(10), 826-829 (1999-09-24)
Effects of orchiectomy on male Calomys callosus infected with the Y strain of Trypanosoma cruzi were studied. Male C. callosus of the same age and weight were divided into three groups: intact, sham operated, and castrated. After 1 month they
H A Verheul et al.
Immunopharmacology, 11(2), 93-99 (1986-04-01)
Nandrolone decanoate injections (20, 40 or 80 mg/kg/3 weeks) in female or castrated male New Zealand black/white mice, starting at various ages (4, 9, 17 or 26 weeks) prolong survival, reduce proteinuria and affect the weights of various endocrine and
G Haidl et al.
Der Urologe. Ausg. A, 47(12), 1555-1556 (2008-11-18)
Evidence-based drug therapy for male infertility is often difficult because 30% of all cases of male infertility are classified as idiopathic, and another 30% need surgical treatment. Without knowledge of the underlying pathology, there is no foundation for a specific
John K Amory
Current opinion in endocrinology, diabetes, and obesity, 15(3), 255-260 (2008-04-29)
Testosterone functions as a contraceptive by suppressing the secretion of luteinizing hormone and follicle-stimulating hormone from the pituitary. Low concentrations of these hormones deprive the testes of the signals required for spermatogenesis and results in markedly decreased sperm concentrations and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service