Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T1132

Sigma-Aldrich

Tetraethylthiuram disulfide

≥97%

Synonym(s):

Bis(diethylthiocarbamoyl) disulfide, Bis(diethylthiocarbamyl) disulfide, Disulfiram, TETD

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)2NCSS2CSN(C2H5)2
CAS Number:
Molecular Weight:
296.54
Beilstein/REAXYS Number:
1712560
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

≥97%

mp

69-71 °C (lit.)

SMILES string

CCN(CC)C(=S)SSC(=S)N(CC)CC

InChI

1S/C10H20N2S4/c1-5-11(6-2)9(13)15-16-10(14)12(7-3)8-4/h5-8H2,1-4H3

InChI key

AUZONCFQVSMFAP-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

Alcohol dehydrogenase inhibitor.
Studies identify disulfiram as a prototype for development of novel cancer preventive agents that target redox-regulatory Cys switches in PKC isozymes. Proceedings of the AACR, Volume 45, March 2004. #2242

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1 - STOT RE 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ghali Brahemi et al.
Journal of medicinal chemistry, 53(7), 2757-2765 (2010-03-13)
The zinc-ejecting aldehyde dehydrogenase (ALDH) inhibitory drug disulfiram (DSF) was found to be a breast cancer-associated protein 2 (BCA2) inhibitor with potent antitumor activity. We herein describe our work in the synthesis and evaluation of new series of zinc-affinic molecules
Beatriz Mothe et al.
The Journal of antimicrobial chemotherapy, 70(6), 1833-1842 (2015-03-01)
The safety, immunogenicity, impact on the latent reservoir and rebound of viral load after therapeutic HIV-1 vaccination with recombinant modified vaccinia Ankara-based (MVA-B) HIV-1 vaccine expressing monomeric gp120 and the fused Gag-Pol-Nef polyprotein of clade B with or without a
Charlotte H Jørgensen et al.
Alcoholism, clinical and experimental research, 35(10), 1749-1758 (2011-05-28)
Alcohol use disorders (AUD) involving hazardous, harmful, and addictive misuse of alcohol are widespread in most parts of the world. The aim of this study was to review the effect of disulfiram in the treatment of patients with AUD. The
Boris Cvek et al.
Drug discovery today, 13(15-16), 716-722 (2008-06-27)
The major approach to the development of anticancer drugs involves searching for new compounds, efficient against malignancies, which are not, as yet, used clinically. This strategy is time-consuming and expensive. Recent studies have disclosed a surprising, but mechanistically consistent, anticancer
Joanna Triscott et al.
Oncotarget, 3(10), 1112-1123 (2012-10-11)
Glioblastomas (GBM) are associated with high rates of relapse. These brain tumors are often resistant to chemotherapies like temozolomide (TMZ) and there are very few treatment options available to patients. We recently reported that polo-like kinase-1 (PLK1) is associated with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service