Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T1033

Sigma-Aldrich

Thymine

suitable for plant cell culture, ~99%

Synonym(s):

2,4-Dihydroxy-5-methylpyrimidine, 5-Methyluracil

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H6N2O2
CAS Number:
Molecular Weight:
126.11
Beilstein/REAXYS Number:
117880
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

assay

~99%

technique(s)

cell culture | plant: suitable

mp

~320 °C (dec.) (lit.)

SMILES string

CC1=CNC(=O)NC1=O

InChI

1S/C5H6N2O2/c1-3-2-6-5(9)7-4(3)8/h2H,1H3,(H2,6,7,8,9)

InChI key

RWQNBRDOKXIBIV-UHFFFAOYSA-N

Gene Information

mouse ... Tymp(72962)

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Supriya Tilvi et al.
Journal of natural products, 73(4), 720-723 (2010-02-20)
The study of the n-butanol extract of the New Caledonian sponge Agelas dendromorpha led to the isolation and identification of three new pyrrole-2-aminoimidazole (P-2-AI) alkaloids, named agelastatins E (3) and F (4) and benzosceptrin C (5), together with 10 known
Manohar Sharma Vedula et al.
Bioorganic & medicinal chemistry, 18(17), 6329-6339 (2010-08-03)
Influenza virus infection constitutes a significant health problem in need of more effective therapies. We have recently identified ((2R,3S,4R,5R)-3-acetoxy-5-(4-benzamido-2-oxopyrimidin-1(2H)-yl)-4-fluoro-3,4-dimethyl-tetrahydrofuran-2-yl) methyl benzoate (18c) as a potent influenza virus inhibitor. We now here report the synthesis and evaluation of a series of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service