Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T0141

Supelco

Testosterone 17β-cypionate

Synonym(s):

17β-Hydroxy-4-androsten-3-one 17-cyclopentylpropionate, 4-Androsten-17β-ol-3-one 17-cypionate, Depotestosterone, Depovirin, Testosterone 17β-cyclopentanepropionate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H40O3
CAS Number:
Molecular Weight:
412.60
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

drug control

USDEA Schedule III; Home Office Schedule 4.2; regulated under CDSA - not available from Sigma-Aldrich Canada

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
veterinary

format

neat

Storage temp.

2-8°C

SMILES string

[H][C@@]12CCC3=CC(=O)CC[C@]3(C)[C@@]1([H])CC[C@]4(C)[C@H](CC[C@@]24[H])OC(=O)CCC5CCCC5

InChI

1S/C27H40O3/c1-26-15-13-20(28)17-19(26)8-9-21-22-10-11-24(27(22,2)16-14-23(21)26)30-25(29)12-7-18-5-3-4-6-18/h17-18,21-24H,3-16H2,1-2H3/t21-,22-,23-,24-,26-,27-/m0/s1

Inchi Key

HPFVBGJFAYZEBE-ZLQWOROUSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Repr. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Becker et al.
The Journal of steroid biochemistry and molecular biology, 125(3-5), 192-201 (2011-01-29)
The application of anabolic steroids in food producing animals is forbidden in the EU since 1988, but the abuse of such drugs is a potential problem. The existing test systems are based on known compounds and can be eluded by
Shannon G Fischer et al.
Behavioural brain research, 180(1), 77-85 (2007-04-10)
Male Syrian hamsters (Mesocricetus auratus) treated with moderately high doses (5.0mg/kg/day) of anabolic/androgenic steroids (AAS) during adolescence (P27-P56) display highly escalated offensive aggression. The current study examined whether adolescent AAS-exposure influenced the immunohistochemical localization of phosphate-activated glutaminase (PAG), the rate-limiting
Jared J Schwartzer et al.
Behavioural pharmacology, 21(4), 314-322 (2010-06-18)
In the Syrian hamster, treatment with anabolic/androgenic steroids (AAS) throughout adolescence increases dopamine and D2 receptor expression in the anterior hypothalamus (AH), a brain region implicated in the control of aggression. D2 receptor antagonists have reduced aggression in various species
Ernani Luis Rhoden et al.
The journal of sexual medicine, 7(1 Pt 1), 277-283 (2010-01-28)
Despite increasing medical interest in testosterone therapy (TTh) for men with testosterone deficiency (TD) there is limited information regarding subjective response rates and acceptability of medium- to long-term TTh in routine clinical practice. To evaluate results in a consecutive series
Ana P S Perez et al.
Microscopy research and technique, 75(8), 1084-1092 (2012-04-11)
There is an increasing variety of endocrine disrupting chemicals (EDCs) either with (anti)estrogenic or (anti)androgenic potential widely present in the environment. These xenosteroids may mimic endogenous steroid hormones disrupting the homeostasis of physiological pathways and leading to several disturbances, especially

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service