Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S7143

Sigma-Aldrich

Sodium thiosulfate pentahydrate

ACS reagent, pH 6.0-8.4 (25 °C, 5% in solution), 99.5-101.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2S2O3 · 5H2O
CAS Number:
Molecular Weight:
248.18
EC Number:
MDL number:
UNSPSC Code:
12352200

grade

ACS reagent

assay

99.5-101.0%

impurities

≤0.002% Nitrogen compounds (as N)
≤0.005% Insoluble matter

pH

6.0-8.4 (25 °C, 5% in solution)

anion traces

sulfate, sulfite (as SO42-): ≤0.1%
sulfide (S2-): passes test (limit about 1 ppm)

SMILES string

O.O.O.O.O.[Na+].[Na+].[O-]S([O-])(=O)=S

InChI

1S/2Na.H2O3S2.5H2O/c;;1-5(2,3)4;;;;;/h;;(H2,1,2,3,4);5*1H2/q2*+1;;;;;;/p-2

InChI key

PODWXQQNRWNDGD-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Resgalla et al.
Chemosphere, 89(1), 102-107 (2012-05-26)
Since the development of the TIE (Toxicity Identification and Evaluation) in 1988 it has been assumed that the capacity of EDTA and sodium thiosulfate to complex some metals, and thus remove their toxicity, can be applied to both freshwater and
Melani Sooriyaarachchi et al.
Metallomics : integrated biometal science, 5(3), 197-207 (2013-02-28)
Cis-Platin (CP) is a remarkably effective Pt-based anticancer drug, but it also exhibits severe toxic side-effects, including nephrotoxicity and ototoxicity. Previous studies conducted with mammalian model organisms have clearly demonstrated that the intravenous administration of N-acetyl-L-cysteine (NAC) or sodium thiosulfate
Brooke Bair et al.
Journal of drugs in dermatology : JDD, 10(9), 1042-1044 (2011-11-05)
Calcinosis cutis is a term used to describe a group of disorders which result in calcium deposits in the skin. These disorders can be separated based on etiology. Sodium thiosulfate has been used to systemically treat calciphylaxis with little to
Michael C Reade et al.
Emergency medicine Australasia : EMA, 24(3), 225-238 (2012-06-08)
Cyanide poisoning is uncommon, but generates interest because of the presumed utility of an antidote immediately available in those areas with a high risk of cyanide exposure. As part of its regular review of guidelines, the Australian Resuscitation Council conducted
Stephanie Chiu et al.
American journal of infection control, 43(11), 1208-1212 (2015-08-10)
The efficacies of disinfection by sodium hypochlorite, accelerated hydrogen peroxide (AHP), and quaternary ammonium compound (QUAT) commonly used in health care facilities were determined using the surrogate viruses murine norovirus (MNV-1) and feline calicivirus (FCV). A virus suspension of known

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service