S6646
Sodium molybdate dihydrate
ACS reagent, pH 7.0-10.5 (25 °C, 5% in solution), 99.5-103.0%
Synonym(s):
Molybdic acid sodium salt dihydrate
About This Item
Recommended Products
grade
ACS reagent
assay
99.5-103.0%
impurities
≤0.005% Insoluble matter
pH
7.0-10.5 (25 °C, 5% in solution)
mp
100 °C (dec.) (lit.)
anion traces
chloride (Cl-): ≤0.005%
phosphate (PO43-): ≤5 ppm
sulfate (SO42-): ≤0.015%
cation traces
Fe: ≤0.001%
NH4+: ≤0.001%
heavy metals (as Pb): ≤5 ppm
SMILES string
O.O.[Na+].[Na+].[O-][Mo]([O-])(=O)=O
InChI
1S/Mo.2Na.2H2O.4O/h;;;2*1H2;;;;/q;2*+1;;;;;2*-1
InChI key
FDEIWTXVNPKYDL-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service