Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S6316

Sigma-Aldrich

Sodium chloride solution

5 M, Biotechnology Performance Certified, suitable for cell culture

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaCl
CAS Number:
Molecular Weight:
58.44
Beilstein/REAXYS Number:
3534976
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

Biotechnology Performance Certified

sterility

0.2 μm filtered

concentration

5 M

technique(s)

cell culture | mammalian: suitable

impurities

Bioburden, tested
DNase, RNase, Protease, tested
endotoxin, tested
≤5 ppm heavy metals (asPb)

SMILES string

[Na+].[Cl-]

InChI

1S/ClH.Na/h1H;/q;+1/p-1

InChI key

FAPWRFPIFSIZLT-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhenghong Xu et al.
Journal of medicinal chemistry, 54(21), 7567-7578 (2011-10-05)
Membrane-associated serine protease matriptase has been implicated in human diseases and might be a drug target. In the present study, a novel class of matriptase inhibitors targeting zymogen activation is developed by a combination of the screening of compound library
Peter Morden et al.
AJR. American journal of roentgenology, 202(1), W13-W18 (2013-12-29)
The purpose of this article is to determine the rate and the cause of displacement of CT power-injectable peripherally inserted central catheters (CT-PICCs) during contrast material and saline flush injection and to modify CT-scanning protocols to decrease the frequency of
Yali V Zhang et al.
Science (New York, N.Y.), 340(6138), 1334-1338 (2013-06-15)
Below a certain level, table salt (NaCl) is beneficial for animals, whereas excessive salt is harmful. However, it remains unclear how low- and high-salt taste perceptions are differentially encoded. We identified a salt-taste coding mechanism in Drosophila melanogaster. Flies use
Anne G M Schilder et al.
Otolaryngology--head and neck surgery : official journal of American Academy of Otolaryngology-Head and Neck Surgery, 149(6), 813-816 (2013-11-22)
The "Cochrane Corner" is a section in the journal that highlights systematic reviews relevant to otolaryngology-head and neck surgery, with invited commentary to aid clinical decision making. This installment features a Cochrane Review, "Interventions for the Prevention of Postoperative Ear
J P Kalyan et al.
The British journal of surgery, 100(13), 1739-1746 (2013-11-15)
Excessive intravenous fluid prescription may play a causal role in postoperative complications following major gastrointestinal resectional surgery. The aim of this study was to investigate whether fluid and salt restriction would decrease postoperative complications compared with a more modern controlled

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service