Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S2882

Sigma-Aldrich

Sodium borodeuteride

Synonym(s):

Sodium tetradeuterioboron

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaBD4
CAS Number:
Molecular Weight:
41.86
EC Number:
MDL number:
UNSPSC Code:
12352103

isotopic purity

98 atom % D

autoignition temp.

752 °F

mass shift

M+4

SMILES string

[Na+].[2H][B-]([2H])([2H])[2H]

InChI

1S/BH4.Na/h1H4;/q-1;+1/i1D4;

InChI key

YOQDYZUWIQVZSF-XWFVQAFUSA-N

Looking for similar products? Visit Product Comparison Guide

Replaced by

Product No.
Description
Pricing

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Skin Corr. 1B - Water-react 1

Storage Class

4.3 - Hazardous materials which set free flammable gases upon contact with water

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service