All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C21H22N2O2 · HNO3
CAS Number:
Molecular Weight:
397.42
EC Number:
MDL number:
UNSPSC Code:
12352000
Recommended Products
SMILES string
O[N+]([O-])=O.[H][C@@]12[C@@H]3CC(=O)N4c5ccccc5[C@@]6(CC[N@H]7CC(=CCO3)[C@@H]1C[C@@H]67)[C@]24[H]
Biochem/physiol Actions
Convulsant; glycine receptor antagonist not associated with the NMDA receptor.
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1
Storage Class
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service