Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S1513

Sigma-Aldrich

Sodium perchlorate

98.0-102.0%

Synonym(s):

Hyperchloric acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaClO4
CAS Number:
Molecular Weight:
122.44
EC Number:
MDL number:
UNSPSC Code:
12352300

assay

98.0-102.0%

reaction suitability

reagent type: oxidant

mp

468 °C (lit.)

SMILES string

[Na+].[O-]Cl(=O)(=O)=O

InChI

1S/ClHO4.Na/c2-1(3,4)5;/h(H,2,3,4,5);/q;+1/p-1

InChI key

BAZAXWOYCMUHIX-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Ox. Sol. 1 - STOT RE 2

target_organs

Thyroid

Storage Class

5.1A - Strongly oxidizing hazardous materials

wgk_germany

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sungnam Park et al.
The journal of physical chemistry. B, 113(22), 7825-7835 (2009-05-14)
The structural and dynamical properties of aqueous ionic solutions influence a wide range of natural and biological processes. In these solutions, water has the opportunity to form hydrogen bonds with other water molecules and anions. Knowing the time scale with
Amir Sabet et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 52(7), 1063-1067 (2011-06-18)
(99m)Tc-macroaggregated albumin ((99m)Tc-MAA) scanning precedes radioembolization of the liver to detect extrahepatic shunting to the lung or gastrointestinal tract. Despite strict preventive measures in the production of (99m)Tc-MAA and in scanning protocols, the images frequently show a gastric concentration of
Heribert Hänscheid et al.
The Journal of clinical endocrinology and metabolism, 96(11), 3511-3516 (2011-08-26)
People being exposed to potentially harmful amounts of radioactive iodine need prophylaxis to prevent high radiation-absorbed doses to the thyroid. Parameters determining the individual protective effect of a pharmacological intervention were investigated. Biokinetics of (123)I was evaluated in 27 healthy
Masatoshi Shibuya et al.
Chemical communications (Cambridge, England), (13)(13), 1739-1741 (2009-03-19)
A facile, green, one-pot oxidation of primary alcohols to carboxylic acids with broad substrate applicability has been developed by employing an expedient catalytic system consisting of 1-Me-AZADO+X-/NaClO2.
Minbiao Ji et al.
The Journal of chemical physics, 134(4), 044516-044516 (2011-02-02)
The dynamics of hydrogen bond (H-bond) formation and dissociation depend intimately on the dynamics of water rotation. We have used polarization resolved ultrafast two-dimensional infrared (2DIR) spectroscopy to investigate the rotational dynamics of deuterated hydroxyl groups (OD) in a solution

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service