Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S1274

Sigma-Aldrich

6-Selenoguanosine

~80%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H13N5O4Se
CAS Number:
Molecular Weight:
346.20
MDL number:
UNSPSC Code:
12352204

assay

~80%

storage temp.

−20°C

SMILES string

NC1=NC(=[Se])c2ncn(C3OC(CO)C(O)C3O)c2N1

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J B Melvin et al.
Cancer research, 44(7), 2794-2798 (1984-07-01)
The effects of 50% lethal doses of three purine analogues, 6-methylmercaptopurine riboside ( MMPR ), 6-thioguanosine (TGR), and 6- selenoguanosine ( SeGR ), on mitotic activity in a slow-growing ( SS1H ) and a fast-growing ( BH3 ) transplantable hepatocellular
A J Repta et al.
Journal of pharmaceutical sciences, 70(6), 634-637 (1981-06-01)
The degradation of 6-selenoguanosine (NSC 137679) (I) in water and in various buffer systems was investigated. Drug degradation in aqueous media was monitored by high-pressure of I in various chromatography. Some kinetic aspects of the degradation of I in various
Y A Beltagy et al.
Journal of pharmaceutical sciences, 69(10), 1168-1170 (1980-10-01)
6-Selenoguanosine (NSC 137679) was stablized and formulated as a lyophilized parenteral paroduct using ascorbic acid as an antioxidant. In addition to preventing the oxidation of 6-selenoguanosine to the corresponding diselenide in aqueous solution, ascorbic acid reduced the diselenide already in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service