Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S110

Sigma-Aldrich

(±)-SKF-83566 hydrochloride

solid

Synonym(s):

(±)-7-Bromo-8-hydroxy-3-methyl-1-phenyl-2,3,4,5-tetrahydro-1H-3-benzazepine hydrochloride, (±)-SCH-24543

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H18BrNO · HCl
CAS Number:
Molecular Weight:
368.70
MDL number:
UNSPSC Code:
12352200

form

solid

storage condition

protect from light

color

light tan

solubility

45% (w/v) aq 2-hydroxypropyl-β-cyclodextrin: ≥14 mg/mL
0.1 M HCl: soluble
H2O: insoluble
ethanol: soluble

SMILES string

Cl.CN1CCc2cc(Br)c(O)cc2C(C1)c3ccccc3

Biochem/physiol Actions

Selective D1 dopamine antagonist; useful in studies involving the radio-brominated derivative.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of a radiobrominated analog of SCH-23390, a selective dopamine D1/DA1 antagonist.
DeJesus, et al.
Journal of Labelled Compounds & Radiopharmaceuticals, 23, 919-919 (1986)
J Arnt et al.
Journal of neural transmission, 67(3-4), 225-240 (1986-01-01)
The effects of 30 dopamine (DA) antagonists, including 4 as stereoisomeric pairs, on circling behaviour induced by the D-1 agonist SKF 38393 and the D-2 agonist pergolide in rats with unilateral 6-hydroxy-DA lesions have been studied. SKF 38393-induced circling was
K E Flaim et al.
Life sciences, 36(15), 1427-1436 (1985-04-15)
Fenoldopam (SKF 82526), a dopamine agonist which exhibits D-1 receptor subtype selectivity, was evaluated as a radioligand for this receptor subtype. In saturation studies in rat striatal membrane preparations, [3H]-fenoldopam appeared to label a single binding site with a KD

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service