Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S0626

Sigma-Aldrich

Sodium persulfate

powder

Synonym(s):

Sodium peroxodisulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2S2O8
CAS Number:
Molecular Weight:
238.10
EC Number:
MDL number:
UNSPSC Code:
12352103

form

powder

color

white

SMILES string

[Na+].[Na+].[O-]S(=O)(=O)OOS([O-])(=O)=O

InChI

1S/2Na.H2O8S2/c;;1-9(2,3)7-8-10(4,5)6/h;;(H,1,2,3)(H,4,5,6)/q2*+1;/p-2

InChI key

CHQMHPLRPQMAMX-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Ox. Sol. 3 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

5.1B - Oxidizing hazardous materials

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chenju Liang et al.
Chemosphere, 70(3), 426-435 (2007-08-19)
In situ chemical oxidation with persulfate anion (S2O82*) is a viable technique for remediation of groundwater contaminants such as trichloroethylene (TCE). An accelerated reaction using S2O82* to destroy TCE can be achieved via chemical activation with ferrous ion to generate
Xiaodan Zhao et al.
Journal of the American Chemical Society, 132(16), 5837-5844 (2010-04-03)
By palladium catalysis, the C-H bond functionalization of O-phenylcarbamates with simple arenes has been achieved using sodium persulfate (Na(2)S(2)O(8)), an inexpensive, easy-to-handle, and environmentally friendly oxidant. This oxidative cross-coupling involves two aromatic C-H bonds undergoing concomitant oxidation to furnish a
Chia-Hsien Yen et al.
Journal of hazardous materials, 186(2-3), 2097-2102 (2011-01-25)
In this study, batch experiments were conducted to evaluate the feasibility of petroleum-hydrocarbon contaminated soil remediation using persulfate oxidation. Various controlling factors including different persulfate and ferrous ion concentrations, different oxidants (persulfate, hydrogen peroxide, and permanganate), and different contaminants (diesel
Ya-Ting Lin et al.
Chemosphere, 82(8), 1168-1172 (2011-01-12)
Using ultraviolet photolytic persulfate activation to produce two sulfate radicals (SO(4)(-)) exhibits a potential for destroying organic contaminants in wastewater treatment applications. This study investigated both the feasibility of using a UV/SPS (sodium persulfate) process to treat phenol in aqueous
Zhiguo Zhang et al.
The Journal of organic chemistry, 77(17), 7665-7670 (2012-08-02)
Mediated by sodium persulfate (Na(2)S(2)O(8)), a series of polysubstituted 4-pyridones were synthesized via self-condensation of N-aryl acetoacetamides, during which a novel N to C 1,3-acyl migration should be involved. The structure of 4-pyridone was unequivocally confirmed by X-ray diffraction analysis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service