Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S0125

Sigma-Aldrich

D-Saccharic acid calcium salt tetrahydrate

Synonym(s):

Calcium D-saccharate tetrahydrate, D-Glucaric acid calcium salt, D-Saccharic acid calcium salt, Calcium D-glucarate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8CaO8 · 4H2O
CAS Number:
Molecular Weight:
320.26
Beilstein/REAXYS Number:
3821334
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:

SMILES string

[H]O[H].[H]O[H].[H]O[H].[H]O[H].O[C@H]1[C@H](O)[C@@H](O)C(=O)O[Ca]OC(=O)[C@H]1O

InChI

1S/C6H10O8.Ca.4H2O/c7-1(3(9)5(11)12)2(8)4(10)6(13)14;;;;;/h1-4,7-10H,(H,11,12)(H,13,14);;4*1H2/q;+2;;;;/p-2/t1-,2-,3-,4+;;;;;/m0...../s1

InChI key

ZQWFSIZRQANUDA-WQMSYZFBSA-L

Application

D-Saccharate (D-glucaric acid) is used to study its potential chemopreventive properties and anti-oxidative properties at the level of peroxynitrite-induced damages.

Biochem/physiol Actions

Calcium glucarate is a constituent of many fruits (apples, citrus fruits) and cruciferous vegetables. Inhibits β-glucuronidase, a phase II enzyme that is elevated in some hormone-dependent cancers of the breast, prostate, and colon.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service