Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

R5380

Sigma-Aldrich

N-Acetyl-Renin Substrate Tetradecapeptide porcine

≥97% (HPLC), suitable for MALDI-MS

Synonym(s):

Ac-Asp-Arg-Val-Tyr-Ile-His-Pro-Phe-His-Leu-Leu-Val-Tyr-Ser

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C87H125N21O21
CAS Number:
Molecular Weight:
1801.05
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Product Name

N-Acetyl-Renin Substrate Tetradecapeptide porcine, ≥97% (HPLC)

Quality Level

assay

≥97% (HPLC)

form

powder

technique(s)

MALDI-MS: suitable

color

white

Storage temp.

−20°C

SMILES string

CCC(C)C(NC(=O)C(Cc1ccc(O)cc1)NC(=O)C(NC(=O)C(CCCNC(N)=N)NC(=O)C(CC(O)=O)NC(C)=O)C(C)C)C(=O)NC(Cc2cnc[nH]2)C(=O)N3CCCC3C(=O)NC(Cc4ccccc4)C(=O)NC(Cc5cnc[nH]5)C(=O)NC(CC(C)C)C(=O)NC(CC(C)C)C(=O)NC(C(C)C)C(=O)NC(Cc6ccc(O)cc6)C(=O)NC(CO)C(O)=O

InChI

1S/C87H125N21O21/c1-12-49(10)72(107-80(122)63(36-53-24-28-57(112)29-25-53)102-82(124)70(47(6)7)105-73(115)58(20-16-30-92-87(88)89)96-78(120)65(39-69(113)114)95-50(11)110)84(126)103-66(38-55-41-91-44-94-55)85(127)108-31-17-21-68(108)81(123)100-61(34-51-18-14-13-15-19-51)75(117)99-64(37-54-40-90-43-93-54)77(119)97-59(32-45(2)3)74(116)98-60(33-46(4)5)79(121)106-71(48(8)9)83(125)101-62(35-52-22-26-56(111)27-23-52)76(118)104-67(42-109)86(128)129/h13-15,18-19,22-29,40-41,43-49,58-68,70-72,109,111-112H,12,16-17,20-21,30-39,42H2,1-11H3,(H,90,93)(H,91,94)(H,95,110)(H,96,120)(H,97,119)(H,98,116)(H,99,117)(H,100,123)(H,101,125)(H,102,124)(H,103,126)(H,104,118)(H,105,115)(H,106,121)(H,107,122)(H,113,114)(H,128,129)(H4,88,89,92)

InChI key

IUJUQXJOVSWPHY-UHFFFAOYSA-N

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jadwiga Dudkiewicz-Wilczyńska et al.
Acta poloniae pharmaceutica, 61(3), 171-174 (2004-10-16)
A high-performance liquid chromatographic assay has been developed to the separation of angiotensin I, tetradecapeptide and the tetrapeptide Leu-Val-Tyr-Ser. This purpose is achieved in a single step, using HPLC technique in the reversed phase system. The method is based on
T C Cheng et al.
Protein science : a publication of the Protein Society, 8(11), 2474-2486 (1999-12-14)
A novel metallocarboxypeptidase (PfuCP) has been purified to homogeneity from the hyperthermophilic archaeon, Pyrococcus furiosus, with its intended use in C-terminal ladder sequencing of proteins and peptides at elevated temperatures. PfuCP was purified in its inactive state by the addition
C F Santos et al.
Journal of periodontology, 80(1), 130-139 (2009-02-21)
The systemic renin-angiotensin system (RAS) promotes the plasmatic production of angiotensin (Ang) II, which acts through interaction with specific receptors. There is growing evidence that local systems in various tissues and organs are capable of generating angiotensins independently of circulating
Khalid M Elased et al.
Hypertension (Dallas, Tex. : 1979), 46(4), 953-959 (2005-08-17)
This article demonstrates the applicability of quantitative proteomics to assays of proteolytic enzyme activity. A novel assay was developed for measurement of renin and angiotensin-converting enzyme (ACE) activity in plasma. The method was validated in animal models associated with alterations

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service