Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P9300

Sigma-Aldrich

Tetraethylene glycol monotetradecyl ether

Synonym(s):

C14E4, Polyoxyethylene (4) myristyl ether, Tetradecyl tetraethylene glycol ether, Tetradecyltetraglycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)13(OCH2CH2)4OH
CAS Number:
Molecular Weight:
390.60
Beilstein/REAXYS Number:
1879291
MDL number:
UNSPSC Code:
12161900

description

non-ionic

mp

30-31 °C

SMILES string

CCCCCCCCCCCCCCOCCOCCOCCOCCO

InChI

1S/C22H46O5/c1-2-3-4-5-6-7-8-9-10-11-12-13-15-24-17-19-26-21-22-27-20-18-25-16-14-23/h23H,2-22H2,1H3

Inchi Key

OARYCGKAYBBHAM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service