Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P9202

Sigma-Aldrich

1,2-Dielaidoyl-sn-glycero-3-phosphocholine

≥98% (GLC, TLC)

Synonym(s):

L-α-Phosphatidylcholine, dielaidoyl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H84NO8P
CAS Number:
Molecular Weight:
786.11
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:

assay

≥98% (GLC, TLC)

functional group

amine

shipped in

ambient

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(CCCCCCC/C=C/CCCCCCCC)=O)COC(CCCCCCC/C=C/CCCCCCCC)=O)=O

InChI

1S/C44H85NO8P/c1-6-8-10-12-14-16-18-20-22-24-26-28-30-32-34-36-43(46)50-40-42(41-52-54(48,49)51-39-38-45(3,4)5)53-44(47)37-35-33-31-29-27-25-23-21-19-17-15-13-11-9-7-2/h20-23,42H,6-19,24-41H2,1-5H3,(H,48,49)/b22-20+,23-21+

InChI key

MWJBWPDYNFAJFV-DQPVQCHKSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N Bezlyepkina et al.
Biophysical journal, 104(7), 1456-1464 (2013-04-09)
We propose a novel, to our knowledge, method for the determination of tie lines in a phase diagram of ternary lipid mixtures. The method was applied to a system consisting of dioleoylphosphatidylcholine (DOPC), egg sphingomyelin (eSM), and cholesterol (Chol). The
Lucie Caillon et al.
Biochimica et biophysica acta, 1828(9), 2091-2098 (2013-05-28)
Human islet amyloid polypeptide (IAPP) forms amyloid fibrils in the pancreatic islets of patients suffering from type 2 diabetes mellitus (T2DM). The formation of IAPP fibrils has been shown to cause membrane damage which most likely is responsible for the
Patrick Walsh et al.
The Journal of biological chemistry, 289(15), 10419-10430 (2014-02-21)
The formation of fibrillar aggregates has long been associated with neurodegenerative disorders such as Alzheimer and Parkinson diseases. Although fibrils are still considered important to the pathology of these disorders, it is now widely understood that smaller amyloid oligomers are
Amanda Musgrove et al.
Langmuir : the ACS journal of surfaces and colloids, 29(10), 3347-3360 (2013-02-19)
The potential-controlled incorporation of DOPC liposomes (100 nm diameter) into an adsorbed octadecanol layer on Au(111) was studied using electrochemical and in situ fluorescence microscopy. The adsorbed layer of octadecanol included a small amount of a lipophilic fluorophore-octadecanol modified with
Marc Torrent et al.
The Biochemical journal, 456(1), 99-108 (2013-08-22)
Vertebrate secreted RNases (ribonucleases) are small proteins that play important roles in RNA metabolism, angiogenesis or host defence. In the present study we describe the antimicrobial properties of the N-terminal domain of the hcRNases (human canonical RNases) and show that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service